APO455835884 Apollo Scientific Ltd

4-Fluoro-3-methoxybenzyl alcohol

  • MDL number MFCD00272133
  • Cas Number : 128495-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-50
Synonym 2-Fluoro-5-(hydroxymethyl)anisole, (4-Fluoro-3-methoxyphenyl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-methoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-methoxybenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart