APO455835902 Apollo Scientific Ltd

2-Fluoro-6-(4-chlorobenzyloxy)benzonitrile

  • MDL number MFCD00068205
  • Cas Number : 175204-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-148
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-4-chlorobenzyloxybenzonitrile

Safety Information

signalword Danger
pcodes P260;P280;P301+P310;P302+P352;P304+P340;P361+P364
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-4-chlorobenzyloxybenzonitrile
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart