APO455835910 Apollo Scientific Ltd

3-Deoxy-3-fluoro-D-glucopyranose

  • MDL number MFCD00063302
  • Cas Number : 7226-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-117
Synonym (3S,4S,5R,6R)-4-Fluoro-6-(hydroxymethyl)tetrahydro-2H-pyran-2,3,5-triol
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1781

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1781
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart