APO455835912 Apollo Scientific Ltd

5-Fluoro-6-hydroxyhydrouracil monohydrate

  • MDL number MFCD00150905
  • Cas Number : 37103-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >130
Synonym 5-Fluoro-6-hydroxydihydropyrimidine-2,4(1H,3H)-dione monohydrate
Purity 97
Flash Point >110
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1789

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1789
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart