APO455835923 Apollo Scientific Ltd

5-Fluoro-4-hydroxyquinazoline

  • MDL number MFCD00173674
  • Molecular Weight : 164.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-242
Synonym 5-Fluoroquinazolin-4-ol, 5-Fluoro-4-hydroxy-1,3-benzodiazine
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-4-hydroxyquinazoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-4-hydroxyquinazoline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart