APO455835933 Apollo Scientific Ltd

5-(Aminomethyl)-2-(trifluoromethyl)pyridine

  • MDL number MFCD01862649
  • Cas Number : 387350-39-2
  • Molecular Weight : 176.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [6-(Trifluoromethyl)pyridin-3-yl]methylamine, 6-(Trifluoromethyl)nicotinylamine
Purity 96
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 62-64/0.4mm
Product Link https://store.apolloscientific.co.uk/product/5-aminomethyl-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminomethyl-2-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart