APO455835943 Apollo Scientific Ltd

3-Fluoro-4-methoxybenzyl alcohol

  • MDL number MFCD00070642
  • Cas Number : 96047-32-4
  • Molecular Weight : 156.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-4-(hydroxymethyl)anisole
Purity 98
Physical Form Liquid-Viscous
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-methoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-methoxybenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart