APO455835944 Apollo Scientific Ltd

2-Fluoro-5-iodotoluene

  • MDL number MFCD00013710
  • Cas Number : 452-68-6
  • Molecular Weight : 236.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Fluoro-4-iodo-2-methylbenzene
Purity 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-87/9mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-iodotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-iodotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart