APO455835946 Apollo Scientific Ltd

3-Fluoro-4-methoxyphenylacetic acid

  • MDL number MFCD00060353
  • Cas Number : 452-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-116
Synonym 4-(Carboxymethyl)-2-fluoroanisole
Purity 98
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-methoxyphenylacetic-acid-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-methoxyphenylacetic-acid-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart