APO455835951 Apollo Scientific Ltd

1,1,2,3,3,3-Hexafluoropropyl 2,2,2-trifluoroethyl ether

  • MDL number MFCD00476468
  • Cas Number : 993-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1,1,4,4,5,6,6,6-Nonafluoro-3-oxahexane
Purity 97
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 72.7
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3647

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3647
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart