APO455835955 Apollo Scientific Ltd

4'-Fluoro-2'-methoxyacetophenone

  • MDL number MFCD00061143
  • Cas Number : 51788-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-methoxyacetophenone-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-methoxyacetophenone-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart