APO455835960 Apollo Scientific Ltd

4-Fluoro-4'-methoxybenzophenone

  • MDL number MFCD00055469
  • Cas Number : 345-89-1
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Synonym (4-Fluorophenyl)(4-methoxyphenyl)methanone, 4-(4-Fluorobenzoyl)anisole
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-4-methoxybenzophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-4-methoxybenzophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart