APO455835976 Apollo Scientific Ltd

5-Fluoro-2-methylbenzoyl chloride

  • MDL number MFCD00236681
  • Cas Number : 21900-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.27
Flash Point 87
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 108/24mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1907

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1907
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart