APO455835978 Apollo Scientific Ltd

2-Fluoro-3-methylbenzyl bromide

  • MDL number MFCD00042470
  • Cas Number : 151412-12-3
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-2-fluoro-3-methylbenzene, alpha-Bromo-2-fluoro-m-xylene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 168/15mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-methylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-methylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart