APO455835979 Apollo Scientific Ltd

5-Fluoro-2-methylphenylhydrazine hydrochloride

  • MDL number MFCD00053032
  • Cas Number : 325-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197(dec.)
Synonym 4-Fluoro-2-hydrazinotoluene hydrochloride, 4-Fluoro-2-hydrazino-1-methylbenzene hydrochloride
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1918

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1918
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart