APO455835984 Apollo Scientific Ltd

5-Fluoro-2-methylphenyl isothiocyanate

  • MDL number MFCD00046800
  • Cas Number : 175205-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-isothiocyanatotoluene, 4-Fluoro-2-isothiocyanato-1-methylbenzene
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 101/20mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1923

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1923
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart