APO455835985 Apollo Scientific Ltd

2-Fluoro-6-[(4-methylphenyl)sulphanyl]benzonitrile

  • MDL number MFCD00068207
  • Cas Number : 175204-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 2-Cyano-3-fluoro-4'-methyldiphenyl thioether, 2-Fluoro-6-[(4-methylphenyl)thio]benzonitrile
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1924

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1924
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart