APO455835993 Apollo Scientific Ltd

1,2-Dibromopentafluoropropyl 2,2,2-trifluoroethyl ether

  • MDL number MFCD03094229
  • Cas Number : 396716-52-2
  • Molecular Weight : 389.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Dibromo-4-oxa-5H,5H-octafluorohexane
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/12-dibromopentafluoropropyl-222-trifluoroethyl-ether

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/12-dibromopentafluoropropyl-222-trifluoroethyl-ether
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart