APO455835998 Apollo Scientific Ltd

Methyl 3,3,3-trifluoro-DL-lactate

  • MDL number MFCD02093316
  • Cas Number : 93496-85-6
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Synonym Methyl 2-hydroxy-3,3,3-trifluoropropanoate, Methyl 2-hydroxy-3,3,3-trifluoropropionate
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60-64/20mm
Product Link https://store.apolloscientific.co.uk/product/methyl-333-trifluoro-dl-lactate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-333-trifluoro-dl-lactate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart