APO455836009 Apollo Scientific Ltd

3-Bromophenyl trifluoromethyl sulphide

  • MDL number MFCD00236348
  • Cas Number : 2252-45-1
  • Molecular Weight : 257.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[Trifluoromethyl)thio]bromobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 192-194
Product Link https://store.apolloscientific.co.uk/product/3-bromophenyl-trifluoromethyl-sulphide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromophenyl-trifluoromethyl-sulphide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart