APO455836013 Apollo Scientific Ltd

Trifluoroacetamidine

  • MDL number MFCD00041545
  • Cas Number : 354-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,2-Trifluoroethanimidamide
Storage temp. Store at -20
Shipping Temperature Ship on Ice
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4959

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=4959
Availability details for
Quantity
  • Availability
  • Add to Cart