APO455836023 Apollo Scientific Ltd

5-Bromo-2,4-difluorobenzenesulphonyl chloride

  • MDL number MFCD01940424
  • Cas Number : 287172-61-6
  • Molecular Weight : 291.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-36
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-difluorobenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-difluorobenzenesulphonyl-chloride
pictograms GHS05
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart