APO455836030 Apollo Scientific Ltd

4-Fluoro-4'-nitrobenzophenone

  • MDL number MFCD00053428
  • Cas Number : 2195-47-3
  • Molecular Weight : 245.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89
Purity 98
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-4-nitrobenzophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-4-nitrobenzophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart