APO455836046 Apollo Scientific Ltd

3-Amino-4-nitrobenzotrifluoride

  • MDL number MFCD00042447
  • Cas Number : 402-14-2
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 98-102
Iupac Name 2-nitro-5-(trifluoromethyl)aniline
Inchi Code 1S/C7H5F3N2O2/c8-7(9,10)4-1-2-6(12(13)14)5(11)3-4/h1-3H,11H2
Inchi Key AUTLVHYEAAAKNM-UHFFFAOYSA-N

Description

Synonym 2-Nitro-5-(trifluoromethyl)aniline, 2-Amino-4-(trifluoromethyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 98-102
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-nitrobenzotrifluoride
pictograms GHS06
hstatements H302+H332;H311;H319
Availability details for
Quantity
  • Availability
  • Add to Cart