APO455836047 Apollo Scientific Ltd

4-Benzyloxy-2-fluorobenzeneboronic acid

  • MDL number MFCD02093064
  • Cas Number : 166744-78-1
  • Molecular Weight : 246.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-155
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxy-2-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxy-2-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart