APO455836048 Apollo Scientific Ltd

4-Benzyloxy-3-fluorobenzeneboronic acid

  • MDL number MFCD00994627
  • Cas Number : 133057-83-7
  • Molecular Weight : 246.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-131
Synonym [(4-Borono-2-fluorophenoxy)methyl]benzene
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxy-3-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxy-3-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart