APO455836051 Apollo Scientific Ltd

Bis(3-fluorophenyl) disulphide

  • MDL number MFCD01318104
  • Cas Number : 63930-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,1'-Disulphanediylbis(3-fluorobenzene), 1,1'-Dithiobis(3-fluorobenzene)
Purity 97
Density 1.27
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 308
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2359

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2359
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart