APO455836054 Apollo Scientific Ltd

1,3-Bis(trifluoromethyl)-4-chlorobenzene

  • MDL number MFCD01631638
  • Cas Number : 327-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Bis(trifluoromethyl)chlorobenzene
Purity 97
Density 1.52
Flash Point 59
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 150-154
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2377

Safety Information

signalword Warning
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2377
pictograms GHS02;GHS07
hstatements H226;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart