APO455836064 Apollo Scientific Ltd

3,6-Difluoropyridazine

  • MDL number MFCD09909693
  • Cas Number : 33097-39-1
  • Molecular Weight : 116.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,6-Difluoro-1,2-diazine
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-difluoropyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/36-difluoropyridazine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart