APO455836069 Apollo Scientific Ltd

4-(Dimethylamino)-3-(trifluoroacetyl)pyridine

  • MDL number MFCD09904820
  • Cas Number : 230305-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[4-(Dimethylamino)pyridin-3-yl]-2,2,2-trifluoroethan-1-one, 4-(Dimethylamino)-3-(2,2,2-trifluoroethanoyl)pyridine
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 70/4mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43817

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43817
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart