APO455836071 Apollo Scientific Ltd

4-[(Trifluoromethyl)thio]benzamide

  • MDL number MFCD02689728
  • Cas Number : 330-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-130
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5827

Safety Information

signalword Warning
pcodes P280;P302;P352;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5827
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart