APO455836073 Apollo Scientific Ltd

2-(Trifluoromethyl)benzhydrazide

  • MDL number MFCD00221473
  • Cas Number : 344-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Synonym 2-(Trifluoromethyl)benzoic acid hydrazide
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylbenzhydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylbenzhydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart