APO455836085 Apollo Scientific Ltd

4-Fluoro-3-nitrotoluene

  • MDL number MFCD00007060
  • Cas Number : 446-11-7
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 22-30
Iupac Name 1-fluoro-4-methyl-2-nitrobenzene
density 1.262
Inchi Code 1S/C7H6FNO2/c1-5-2-3-6(8)7(4-5)9(10)11/h2-4H,1H3
Inchi Key OORBDHOQLZRIQR-UHFFFAOYSA-N

Description

Synonym 2-Fluoro-5-methylnitrobenzene, 1-Fluoro-4-methyl-2-nitrobenzene
Purity 99%
Density 1.262
Flash Point 241(dec.)
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 22-30
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-nitrotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-nitrotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart