APO455836089 Apollo Scientific Ltd

1-Hydroxy-6-(trifluoromethyl)-1H-benzotriazole

  • MDL number MFCD00038806
  • Cas Number : 26198-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Synonym 6-(Trifluoromethyl)-1H-benzotriazol-1-ol
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43826

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43826
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart