APO455836103 Apollo Scientific Ltd

2-Chloro-4-fluorobenzyl chloride

  • MDL number MFCD00236026
  • Cas Number : 93286-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-1-(chloromethyl)-4-fluorobenzene, alpha,2-Dichloro-4-fluorotoluene
Purity 96
Density 1.37
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2654

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2654
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart