APO455836108 Apollo Scientific Ltd

2-Chloro-4-fluorophenylacetonitrile

  • MDL number MFCD00236029
  • Cas Number : 75279-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-69
Synonym 2-Chloro-4-fluorobenzyl cyanide
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluorophenylacetonitrile-97

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluorophenylacetonitrile-97
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart