APO455836109 Apollo Scientific Ltd

3-Fluoro-4-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD00236303
  • Cas Number : 238754-67-1
  • Molecular Weight : 222.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-90
Synonym 4-(Carboxymethyl)-2-fluorobenzotrifluoride, [3-Fluoro-4-(trifluoromethyl)phenyl]acetic acid
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethylphenylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-trifluoromethylphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart