APO455836112 Apollo Scientific Ltd

2-Fluoro-6-phenoxybenzonitrile

  • MDL number MFCD00068201
  • Cas Number : 175204-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-93
Synonym 2-Cyano-3-fluorodiphenyl ether
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1995

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1995
pictograms GHS07
hstatements H302;H312;H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart