APO455836116 Apollo Scientific Ltd

2-(Bromomethyl)-4-fluorobenzonitrile

  • MDL number MFCD08059542
  • Cas Number : 421552-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-5-fluorobenzyl bromide, alpha-Bromo-4-fluoro-o-tolunitrile
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10507

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10507
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart