APO455836118 Apollo Scientific Ltd

2,3-Difluoro-6-methoxybenzaldehyde

  • MDL number MFCD02093968
  • Cas Number : 187543-87-9
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-62
Synonym 3,4-Difluoro-2-formylanisole, 5,6-Difluoro-o-anisaldehyde
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-6-methoxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-6-methoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart