APO455836168 Apollo Scientific Ltd

3'-Fluoropropiophenone

  • MDL number MFCD00061245
  • Cas Number : 455-67-4
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3-Fluorophenyl)propan-1-one
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 94-96/5mm
Product Link https://store.apolloscientific.co.uk/product/3-fluoropropiophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoropropiophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart