APO455836180 Apollo Scientific Ltd

5-Amino-3-(4-fluorophenyl)isoxazole

  • MDL number MFCD03407398
  • Cas Number : 81465-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-103
Synonym 3-(4-Fluorophenyl)isoxazol-5-amine
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10476

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10476
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart