APO455836196 Apollo Scientific Ltd

4-Chloro-2-(trifluoromethyl)benzenesulphonyl chloride

  • MDL number MFCD04638751
  • Cas Number : 54090-42-5
  • Molecular Weight : 279.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-(trifluoromethyl)benzenesulfonyl chloride
Inchi Code 1S/C7H3Cl2F3O2S/c8-4-1-2-6(15(9,13)14)5(3-4)7(10,11)12/h1-3H
Inchi Key DQKPVVIVMURTDM-UHFFFAOYSA-N

Description

Synonym 5-Chloro-2-(chlorosulphonyl)benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-trifluoromethylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-trifluoromethylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart