APO455836199 Apollo Scientific Ltd

tert-Butyl 2-(trifluoromethyl)acrylate

  • MDL number MFCD04038275
  • Cas Number : 105935-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym TFMA-B, tert-Butyl 2-(trifluoromethyl)prop-2-enoate
Purity 98
Flash Point >110
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 120
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8240

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8240
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart