APO455836204 Apollo Scientific Ltd

4-(Difluoromethoxy)benzylamine

  • MDL number MFCD00236231
  • Cas Number : 177842-14-7
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [4-(Difluoromethoxy)phenyl]methylamine
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxybenzylamine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxybenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart