APO455836214 Apollo Scientific Ltd

3,4-Difluorobenzyl chloride

  • MDL number MFCD00236218
  • Cas Number : 698-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha-Chloro-3,4-difluorotoluene, 4-(Chloromethyl)-1,2-difluorobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 27-29/0.4mm
Product Link https://store.apolloscientific.co.uk/product/34-difluorobenzyl-chloride-98

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-difluorobenzyl-chloride-98
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart