APO455836224 Apollo Scientific Ltd

2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone

  • MDL number MFCD00061276
  • Cas Number : 74853-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,2-Trifluoro-1-[4-(trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoroacetyl)benzotrifluoride
Purity 97
Density 1.433
Flash Point 56
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 63-65/15mm
Product Link https://store.apolloscientific.co.uk/product/222-trifluoro-4-trifluoromethylacetophenone-97

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/222-trifluoro-4-trifluoromethylacetophenone-97
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart