APO455836226 Apollo Scientific Ltd

Methyl 4-fluoro-3-nitrobenzoate

  • MDL number MFCD00196161
  • Cas Number : 329-59-9
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Synonym 2-Fluoro-5-(methoxycarbonyl)nitrobenzene
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 109/1mm
Product Link https://store.apolloscientific.co.uk/product/methyl-4-fluoro-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-fluoro-3-nitrobenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart