APO455836227 Apollo Scientific Ltd

3,4-Difluorophenacyl chloride

  • MDL number MFCD03966888
  • Cas Number : 51336-95-9
  • Molecular Weight : 190.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-1-(3,4-difluorophenyl)ethan-1-one
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-difluorophenacyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-difluorophenacyl-chloride
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart