APO455836228 Apollo Scientific Ltd

2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid

  • MDL number MFCD00792417
  • Cas Number : 656-46-2
  • Molecular Weight : 202.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-157
Synonym 5-Carboxy-2,2-difluoro-1,3-benzodioxole, 3,4-(Difluoromethylenedioxy)benzoic acid
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/22-difluoro-13-benzodioxole-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22-difluoro-13-benzodioxole-5-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart